missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CEACAM19 Monoclonal Antibody (1032215)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA562022
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
The specific function of the protein remains unknown.
Specifications
| CEACAM19 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q7Z692 | |
| CEACAM19 | |
| Human embryonic kidney cell HEK293-derived human CEACAM-19, Ala33-Gly157. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry | |
| 1032215 | |
| Unconjugated | |
| CEACAM19 | |
| C130022P09Rik; carcinoembryonic antigen related cell adhesion molecule 19; carcinoembryonic antigen-like 1; Carcinoembryonic antigen-related cell adhesion molecule 19; CEACAM19; CEACM19; CEAL1; CEA-related cell adhesion molecule 19; UNQ2973/PRO7436 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 56971 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction