missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEACAM8/CD66b Antibody (6/40c) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
214.00€ - 509.00€
Specifications
| Antigen | CEACAM8/CD66b |
|---|---|
| Clone | 6/40c |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:100 - 1:500, Immunoprecipitation 1:10 - 1:500, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18673838
|
Novus Biologicals
NBP2-54627-0.025mg |
0.025 mg |
214.00€
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18620558
|
Novus Biologicals
NBP2-54627 |
0.1 mg |
509.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CEACAM8/CD66b Monoclonal antibody specifically detects CEACAM8/CD66b in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, CyTOFSpecifications
| CEACAM8/CD66b | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| PBS | |
| 1088 | |
| IgG1 κ | |
| Protein A purified |
| 6/40c | |
| Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:100 - 1:500, Immunoprecipitation 1:10 - 1:500, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| anti human cd66b apc, Carcinoembryonic antigen CGM6, carcinoembryonic antigen gene family member 6, carcinoembryonic antigen-related cell adhesion molecule 8, CD66b, CD66b antigen, cd66b clone g10f5, cd66b G10F5, cd66b granulocyte marker, cd66b ihc, cd66b marker, cd66b neutrophil marker, cd66b neutrophils, CD67, CD67 antigen, CGM6, NCA-95, Non-specific cross-reacting antigen NCA-95 | |
| Recombinant soluble human CEACAM8-Fc produced in HEK293 cells | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -70°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title