missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEACAM8/CD66b Antibody (80H3), Novus Biologicals™
Mouse Monoclonal Antibody
206.00€ - 485.00€
Specifications
| Antigen | CEACAM8/CD66b |
|---|---|
| Clone | 80H3 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:25-1:100 |
| Applications | Western Blot, Immunohistochemistry (Frozen), Immunohistochemistry, Flow Cytometry |
Description
CEACAM8/CD66b Monoclonal specifically detects CEACAM8/CD66b in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
| CEACAM8/CD66b | |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry (Frozen), Immunohistochemistry, Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Carcinoembryonic antigen CGM6, carcinoembryonic antigen gene family member 6, carcinoembryonic antigen-related cell adhesion molecule 8, CD66b, CD66b antigen, CD67, CD67 antigen, CGM6, NCA-95, Non-specific cross-reacting antigen NCA-95 | |
| CEACAM8 | |
| IgG1 | |
| Protein A purified | |
| NB100-64916 reacts with the CGM6 gene product of the carcinoembryonic gene family. This is a 95-100kD GPI anchored molecule. CEACAM8 is expressed strongly by mature granulocytes and metamyelocytes but weakly by bone marrow myelocytes. CEACAM8 is not expressed by peripheral blood monocytes or lymphocytes. |
| 80H3 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:25-1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| P31997 | |
| 1088 | |
| CML leucocytes | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title