Learn More
Cefotiam dihydrochloride, Thermo Scientific Chemicals
Product Code. 15467929
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15467929 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15467929 Supplier Thermo Scientific Alfa Aesar Supplier No. J66529.MC
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
missing translation for 'validMainframeMsg'
A broad spectrum antibiotic
A broad spectrum antibiotic
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 66309-69-1
Molecular Formula C18H27Cl2N9O5S3
Molecular Weight (g/mol) 616.552
MDL Number MFCD09878605
InChI Key KHUCCKUOJMXYKH-BUWDDJPCSA-N
PubChem CID 70590037
IUPAC Name (6R,7R)-7-[[2-(2-amino-1,3-thiazol-4-yl)acetyl]amino]-3-[[1-[2-(dimethylamino)ethyl]tetrazol-5-yl]sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydrate;dihydrochloride
SMILES CN(C)CCN1C(=NN=N1)SCC2=C(N3C(C(C3=O)NC(=O)CC4=CSC(=N4)N)SC2)C(=O)O.O.Cl.Cl
Specifications
Type Cefotiam Dihydrochloride
Physical Form Powder
Color White
Quantity 100 mg
Flash Point >110°C (230°F)
Boiling Point 94°C
Solubility Information Soluble in water
Formula Weight 562.09
Chemical Name or Material Cefotiam dihydrochloride