missing translation for 'onlineSavingsMsg'
Learn More

Cefotiam Dihydrochloride, TRC

Product Code. 30137774
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137774 100 mg 100mg
30134685 250 mg 250mg
17952235 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137774 Supplier Toronto Research Chemicals Supplier No. TRCC243005100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66309-69-1
Molecular Formula C18 H23 N9 O4 S3 . 2[HCl]
Molecular Weight (g/mol) 598.55
Synonym 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[2-(2-amino-4-thiazolyl)acetyl]amino]-3-[[[1-[2-(dimethylamino)ethyl]-1H-tetrazol-5-yl]thio]methyl]-8-oxo-, hydrochloride (1:2), (6R,7R)-, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2-amino-4-thiazolyl)acetyl]amino]-3-[[[1-[2-(dimethylamino)ethyl]-1H-tetrazol-5-yl]thio]methyl]-8-oxo-, dihydrochloride, (6R,7R)- (9CI), 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2-amino-4-thiazolyl)acetyl]amino]-3-[[[1-[2-(dimethylamino)ethyl]-1H-tetrazol-5-yl]thio]methyl]-8-oxo-, dihydrochloride, (6R-trans)-, Abbott 48999, Cefotiam dihydrochloride, Cefotiam hydrochloride, Ceradon, HAR, HNP, Halospor, KMS, PAS, Pansporin, Pansporine, SEP, SFC, SFP, Spizef, Sporidyn
IUPAC Name (6R,7R)-7-[[2-(2-amino-1,3-thiazol-4-yl)acetyl]amino]-3-[[1-[2-(dimethylamino)ethyl]tetrazol-5-yl]sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;dihydrochloride
SMILES Cl.Cl.CN(C)CCn1nnnc1SCC2=C(N3[C@H](SC2)[C@H](NC(=O)Cc4csc(N)n4)C3=O)C(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C18H23N9O4S3.2ClH/c1-25(2)3-4-26-18(22-23-24-26)34-7-9-6-32-15-12(14(29)27(15)13(9)16(30)31)21-11(28)5-10-8-33-17(19)20-10;;/h8,12,15H,3-7H2,1-2H3,(H2,19,20)(H,21,28)(H,30,31);2*1H/t12-,15-;;/m1../s1
Formula Weight 597.0569
Percent Purity >95%
Chemical Name or Material Cefotiam HCl
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.