missing translation for 'onlineSavingsMsg'
Learn More

Cefoxitin sodium, 92.7-97%, Thermo Scientific™

A broad spectrum antibiotic

Brand:  Thermo Scientific Alfa Aesar J66800.MD

Quantity 250mg

 View more versions of this product

Product Code. 15408309

Request Bulk or Custom Format

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

33564-30-6
449.428
GNWUOVJNSFPWDD-XMZRARIVSA-M
CHEBI:3501
COC1(C2N(C1=O)C(=C(CS2)COC(=O)N)C(=O)[O-])NC(=O)CC3=CC=CS3.[Na+]
C16H16N3NaO7S2
MFCD00079042
23667300
sodium;(6R,7S)-3-(carbamoyloxymethyl)-7-methoxy-8-oxo-7-[(2-thiophen-2-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate

Specifications

33564-30-6
250mg
C16H16N3NaO7S2
843°C
Soluble in water or methanol
COC1(C2N(C1=O)C(=C(CS2)COC(=O)N)C(=O)[O-])NC(=O)CC3=CC=CS3.[Na+]
449.428
CHEBI:3501
92.7 to 97%
White
>110°C (230°F)
MFCD00079042
14,1936
GNWUOVJNSFPWDD-XMZRARIVSA-M
sodium;(6R,7S)-3-(carbamoyloxymethyl)-7-methoxy-8-oxo-7-[(2-thiophen-2-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
23667300
449.43
Cefoxitin sodium
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.