missing translation for 'onlineSavingsMsg'
Learn More

Cefoxitin sodium, 92.7-97%, Thermo Scientific™

A broad spectrum antibiotic

60.90€ - 187.01€

Chemical Identifiers

CAS 33564-30-6
Molecular Formula C16H16N3NaO7S2
Molecular Weight (g/mol) 449.428
MDL Number MFCD00079042
InChI Key GNWUOVJNSFPWDD-XMZRARIVSA-M
PubChem CID 23667300
ChEBI CHEBI:3501
IUPAC Name sodium;(6R,7S)-3-(carbamoyloxymethyl)-7-methoxy-8-oxo-7-[(2-thiophen-2-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
SMILES COC1(C2N(C1=O)C(=C(CS2)COC(=O)N)C(=O)[O-])NC(=O)CC3=CC=CS3.[Na+]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15408309
SDS View Documents
Thermo Scientific Alfa Aesar
J66800.MD
250mg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15498299
SDS View Documents
Thermo Scientific Alfa Aesar
J66800.03
1g
ONLINE EXCLUSIVE PRICE
191.00€
187.01€ / 1g
Save 3.99€
2% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

33564-30-6
449.428
GNWUOVJNSFPWDD-XMZRARIVSA-M
CHEBI:3501
COC1(C2N(C1=O)C(=C(CS2)COC(=O)N)C(=O)[O-])NC(=O)CC3=CC=CS3.[Na+]
C16H16N3NaO7S2
MFCD00079042
23667300
sodium;(6R,7S)-3-(carbamoyloxymethyl)-7-methoxy-8-oxo-7-[(2-thiophen-2-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate

Specifications

33564-30-6
>110°C (230°F)
MFCD00079042
14,1936
GNWUOVJNSFPWDD-XMZRARIVSA-M
sodium;(6R,7S)-3-(carbamoyloxymethyl)-7-methoxy-8-oxo-7-[(2-thiophen-2-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
23667300
449.43
Cefoxitin sodium
White
C16H16N3NaO7S2
843°C
Soluble in water or methanol
COC1(C2N(C1=O)C(=C(CS2)COC(=O)N)C(=O)[O-])NC(=O)CC3=CC=CS3.[Na+]
449.428
CHEBI:3501
92.7 to 97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H317
May cause an allergic skin reaction.

P261-P272-P280g-P302+P352-P333+P313-P363-P501c

H317

missing translation for 'einecsNumber' : 251-574-6

missing translation for 'rtecsNumber' : XI0330500

missing translation for 'tsca' : No

missing translation for 'storageNote' : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.