missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cefoxitin Sodium, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Cefoxitin Sodium
Specifications
Specifications
| Chemical Name or Material | Cefoxitin sodium |
| CAS | 33564-30-6 |
| Analyte or Component Names | Cefoxitin sodium |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C16 H16 N3 O7 S2 . Na |
| Certifications/Compliance | ISO 17034 |
| Synonym | Cefoxitin Sodium, Sodium (6R,7S)-3-[(carbamoyloxy)methyl]-7-methoxy-8-oxo-7-[[(thiophen-2-yl)acetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 3-[[(aminocarbonyl)oxy]methyl]-7-methoxy-8-oxo-7-[(2-thienylacetyl)amino]-, monosodium salt, (6R,7S)- (9CI), 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 3-[[(aminocarbonyl)oxy]methyl]-7-methoxy-8-oxo-7-[(2-thienylacetyl)amino]-, monosodium salt, (6R-cis)-, Betacef, Cefaxilin sodium, Cefoxitin sodium, Cefoxitin sodium salt, Cenomycin, Farmoxin, MK 306, Mefoxin, Mefoxithin, Mefoxitin, Merxin, Monosodium cefoxitin, Sodium cefoxitin |
| InChI Formula | InChI=1S/C16H17N3O7S2.Na/c1-25-16(18-10(20)5-9-3-2-4-27-9)13(23)19-11(12(21)22)8(6-26-15(17)24)7-28-14(16)19;/h2-4,14H,5-7H2,1H3,(H2,17,24)(H,18,20)(H,21,22);/q;+1/p-1/t14-,16+;/m1./s1 |
| SMILES | [Na+].CO[C@]1(NC(=O)Cc2cccs2)[C@H]3SCC(=C(N3C1=O)C(=O)[O-])COC(=O)N |
| IUPAC Name | sodium;(6R,7S)-3-(carbamoyloxymethyl)-7-methoxy-8-oxo-7-[(2-thiophen-2-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction