missing translation for 'onlineSavingsMsg'
Learn More

Ceftibuten Hydrate, TRC

Product Code. 17923395
Change view
Click to view available options
Quantity:
10 mg
100 mg
5 g
Unit Size:
100mg
10mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17923395 5 g 5g
17984375 10 mg 10mg
17913595 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17923395 Supplier Toronto Research Chemicals Supplier No. TRCC2445005G
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118081-37-1
Molecular Formula C15 H14 N4 O6 S2 . H2O
Molecular Weight (g/mol) 410.4
Synonym [6R-[6α,7β(Z)]]-7-[[2-(2-Amino-4-thiazolyl)-4-carboxy-1-oxo-2-butenyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid Hydrate
IUPAC Name (6R,7R)-7-[[(E)-2-(2-aminothiazol-5-yl)-4-carboxy-but-2-enoyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydrate
SMILES O.Nc1ncc(s1)\C(=C\CC(=O)O)\C(=O)N[C@H]2[C@H]3SCC=C(N3C2=O)C(=O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C15H14N4O6S2.H2O/c16-15-17-5-8(27-15)6(1-2-9(20)21)11(22)18-10-12(23)19-7(14(24)25)3-4-26-13(10)19;/h1,3,5,10,13H,2,4H2,(H2,16,17)(H,18,22)(H,20,21)(H,24,25);1H2/b6-1-;/t10-,13-;/m1./s1
Percent Purity >95%
Chemical Name or Material Ceftibuten Hydrate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.