missing translation for 'onlineSavingsMsg'
Learn More

Celecoxib Carboxylic Acid, TRC

Product Code. 30134876
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134876 25 mg 25mg
17901945 10 mg 10mg
17984945 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134876 Supplier Toronto Research Chemicals Supplier No. TRCC25101025MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 170571-01-4
Molecular Formula C17 H12 F3 N3 O4 S
Molecular Weight (g/mol) 411.36
Synonym 4-[1-[4-(Aminosulfonyl)phenyl]-3-(trifluoromethyl)-1H-pyrazol-5-yl]benzoic Acid, Celecoxib Carboxylic Acid
IUPAC Name 4-[2-(4-sulfamoylphenyl)-5-(trifluoromethyl)pyrazol-3-yl]benzoic acid
SMILES NS(=O)(=O)c1ccc(cc1)n2nc(cc2c3ccc(cc3)C(=O)O)C(F)(F)F

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C17H12F3N3O4S/c18-17(19,20)15-9-14(10-1-3-11(4-2-10)16(24)25)23(22-15)12-5-7-13(8-6-12)28(21,26)27/h1-9H,(H,24,25)(H2,21,26,27)
Formula Weight 411.0501
Percent Purity >95%
Chemical Name or Material Celecoxib Carboxylic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.