Learn More
Biochrom Micro-Volume Cell Holder
Product Code. 11300392
Change view
Click to view available options
Type:
Cell Holder
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
11300392 Cell Holder Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11300392 Supplier Biochrom Supplier No. 80210609
Only null left
Add to basket
Add to basket
Micro-Volume Cell Holder is for use with 50 μL cuvettes. Accessory for the Ultrospec 7500 and 2100 Pro Spectrophotometer.
  • Micro-volume cell holder
  • For the Ultrospec 2100 Pro Spectrophotometer
  • Use with 50 μL cuvette (80-2076-38)
Specifications
Compatibility Ultrospec 2100 Pro, 3100, 3300, 4300, 5300, and 6300 Models
Description Cell Holder
Type Cell Holder
Disposable Reusable
For Use With (Equipment) Ultrospec 7500, 2100 Pro, 3100, 3300, 4300, 5300, and 6300 Models
Shape Round