Learn More
Lonza™ Cell Line Nucleofector™ Kit R
Specifications
Specifications
| Efficiency | 99% Transfection efficiency with siRNA duplexes even in suspension cells. |
| For Use With (Equipment) | Nucleofector™ Device |
| Includes | Five different Cell Line Nucleofector™ Solutions: C, L, R, T, and V |
| No. of Reactions | 25 |
| Product Type | Cell Line Nucleofector Kit R |
| Quantity | 1 Ea. |