Learn More
Greiner Bio-One CELLCOAT™ Poly-L-Lysine Protein Coated Flask
Ideal for low-serum, serum-free cultivation and cell differentiation. Greiner Bio-One CELLCOAT™ CELLCOAT™ Poly-L-Lysine Protein Coated Flask helps in promotion of cell adhesion and proliferation.
Brand: Greiner Bio-One 658930
Additional Details : Weight : 0.80000kg
Description
Poly-L-Lysine (PLL) are synthetic molecules which increase cell adhesion to vessels by altering the net charge of the surface without stimulating biological activity.- Coated with Poly-L-Lysine synthetic protein (30-70 kDa)
- For promotion of cell adhesion, proliferation and growth of transfected cell lines, neuronal cell lines, primary neurons and glial cells
- Ready-to-use products
- Includes lid with condensation rings
Specifications
Canted | |
Non-sterile | |
250 mL |
Poly-L-Lysine | |
Filter Screw Cap |