Learn More
Greiner Bio-One CELLSTAR Cell Culture Flasks
GREENER_CHOICE
Product Code. 10044220
Click to view available options
Capacity (Metric):
50 mL
250 mL
550 mL
650 mL
Culture Area:
175 cm2
25 cm2
75 cm2
Closure Type:
Filter Screw Cap
Screw Cap with Vent Position
Includes:
Graduations; writing area
Printed and engraved graduations
Printed and engraved graduations; measuring grid
Printed graduations; labeling field on side
Printed graduations; labeling field on the side
Printed graduations; writing area
Unit Size:
Pack of 120
Pack of 200
Pack of 40
Pack of 50
Product Code. 10044220
Brand: Greiner Bio-One 690170
Only null left
Add to basket
Add to basket
Manufactured from high-grade polystyrene physically surface-treated to improve cell adhesion and proliferation. Greiner Bio-One™ CELLSTAR Cell Culture Flasks are designed to efficiently reach the cell lawn with a cell scraper and provide optimal access with a pipet.
  • Improved cell adhesion through physical surface treatment
  • Neck of high profile flask reduces the risk of wetting the inside of the cap with medium;  provides additional protection against contamination
  • Canted neck
  • TC surface treatment
  • Graduation on the side
  • Stackable
  • Measuring grid
  • Red standard screw cap
  • Sterile and user-friendly packaging
  • Free of detectable DNase, RNase, human DNA, non-pyrogenic
  • Non-Cytotoxic 
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Red
Closure Material HDPE
Capacity (Metric) 50 mL
Material Polystyrene
Includes Printed and engraved graduations; measuring grid
Neck Style Canted
Surface Coating Physical Surface Treatment
Graduated Yes
Sterility Sterile
Closure Type Screw Cap with Vent Position
Culture Area 25 cm2