Learn More
Greiner Bio-One CELLSTAR Cell Culture Flasks
Click to view available options
Capacity (Metric):
50 mL
250 mL
550 mL
650 mL
Culture Area:
175 cm2
25 cm2
75 cm2
Closure Type:
Filter Screw Cap
Screw Cap with Vent Position
Includes:
Graduations; writing area
Printed and engraved graduations
Printed and engraved graduations; measuring grid
Printed graduations; labeling field on side
Printed graduations; labeling field on the side
Printed graduations; writing area
Unit Size:
Pack of 120
Pack of 200
Pack of 40
Pack of 50
Beskrivning
- Improved cell adhesion through physical surface treatment
- Neck of high profile flask reduces the risk of wetting the inside of the cap with medium; provides additional protection against contamination
- Canted neck
- TC surface treatment
- Graduation on the side
- Stackable
- Measuring grid
- Red standard screw cap
- Sterile and user-friendly packaging
- Free of detectable DNase, RNase, human DNA, non-pyrogenic
- Non-Cytotoxic
Specifikationer
Specifikationer
| Closure Color | Red |
| Closure Material | HDPE |
| Capacity (Metric) | 250 mL |
| Material | Polystyrene |
| Includes | Printed graduations; labeling field on the side |
| Neck Style | Canted |
| Surface Coating | Physical Surface Treatment |
| Graduated | Yes |
| Sterility | Sterile |
| Closure Type | Screw Cap with Vent Position |
| Visa mer |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering