Learn More
Eppendorf™ CellXpert™ C170i, Non-Segmented Inner Door, Water-Level Monitoring
Product Code. 16326325
Change view
Click to view available options
Plug Type:
EU
Door Style:
Left-Sided Handle
Right-Sided Handle
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Type Door Style Unit Size
16326325 EU Left-Sided Handle Each
16306325 EU Right-Sided Handle Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Regulatory: This product is under eco-contribution tax.
Product Code. 16326325 Supplier Eppendorf™ Supplier No. 6731000221
 Restricted Item
Estimated Shipment: 03-03-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Regulatory: This product is under eco-contribution tax.
CellXpert™ C170i CO2 incubator provides reliable growth conditions for more reproducible results.
  • Water-level monitoring
  • Advanced user Interface and front USB-ports
  • Fan-less design, smart racking system and innovative insulation concept
  • New magnetic door locking concept for outer and inner doors
  • Seamless chamber, easy disassembling
  • 180°C High Temperature Disinfection (HTD) with protocol, no internal HEPA-filters
  • Customizable tasks and alarms
  • Standard building management system and VisioNize connected lab
  • Vibration and turbulence protection
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 170 L
Chamber Material Stainless Steel
CO2 Sensor Technology IR Sensor
Plug Type EU
Temperature Range (Metric) Ambient +4° to 50°C
Display VisioNize™ touch interface
Door Style Left-Sided Handle
O2 Sensor Technology ZrO2 (zirconium dioxide)
Ports 2
Dimensions (D x W x H) Interior 44.5 x 53.9 x 69.2 cm (17.5 x 21.2 x 27.2 in.)
Dimensions (L x W x H) 71.5 x 71.8 x 90.0 cm (28.3 x 28.1 x 35.4 in.)
Electrical Requirements 220 to 240 V, 50/60 Hz