missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ CellXpert™ CS220 - Large Capacity CO2 Incubator Shaker
GREENER_CHOICE

Product Code. 30213521 Shop All Eppendorf Products
Change view
Click to view available options
Orbit:
25 mm
51 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Orbit unitSize
30213521 25 mm Each
30213523 51 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 30213521 Supplier Eppendorf™ Supplier No. 6740211010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.

Accelerate your research with the Eppendorf™ CellXpert™ CS220 CO2 Incubator Shaker - the only CO2 incubator shaker with integrated 180°C sterilization. Achieve high flask capacity and reliable results with optimized lab space efficiency and low operating costs.

Accelerate your growth - CellXpert™ CS220 - CO2 Incubator Shaker

The CellXpert™ CS220 is a large capacity, stackable CO2 Incubator Shaker that was designed to accelerate mammalian suspension culture research projects.

Please order separately: platform, clamps, sticky pads, storage base or feet, water supply package. Optional: 1-2 stacking kits and water tank holder

  • High flask capacity
  • 180°C sterilization routine integrated: including wizard and downloadable performance protocol, no locally restricted UV-airflow/surface-disinfection
  • Stainless steel chamber: Hygienic design according to EN ISO 14159:2008 standard plus only few internal parts and no drive wheel, crevices, cables or heating fins inside
  • Optimized throughput per lab space: Highest platform size vs. footprint ratio in the market, stackable up to 3-fold
  • Saves costs: Always included: slide-out platform, rH-control, touch interface, access port, 180°C sterilization routine, no UV-lamps that require frequent exchange, low gas and energy consumption
  • Eppendorf X-drive: Proven and tested shaker drive with four weight-supporting shafts for 24/7 vibration-free shaking
  • Glide-up door and slide-out platform: Saves space and ensures fast and easy experiment setup, cleaning and service – even at the back of the chamber
  • Full control and fast root-cause analysis: 7 in. touchscreen shows all setpoints and current parameters at a glance, direct visual overview of logged data/events including door openings and alarms, user management

Applications:

  • Protein Expression
  • Screening
  • Cell line development
  • Early process development
  • Viral vector production in mammalian cells e.g., Ad, AAV, LV
  • Bioreactor inoculum preparation
TRUSTED_SUSTAINABILITY

Specifications

Orbit 25 mm
Rotating Angle
Load Bearing Capacity (Metric) 25 kg
Speed Range 20 to 300 rpm
Voltage 230 V
Temperature Range (Metric) 8°C above ambient to 60°C
Product Type CO2 Incubator Shaker
Load Bearing Capacity (English) 56 lb.
Plug Type EU
Temperature Uniformity ±0.3°C at 37°C
Depth (English) Interior 22.8 in.
Depth (Metric) Interior 58.0 cm
Dimensions (D x W x H) Interior 58.0 x 102.5 x 38.5 cm
Dimensions (LxW) Platform 51.3 x 93 cm
Dimensions (L x W x H) 80.5 x 131.8 x 67.3 cm
Height (English) 26.5 in.
Height (English) Interior 15. 2 in.
Height (Metric) 67.3 cm
Height (Metric) Interior 38.5 cm
Length (English) 31.7 in.
Length (English) Platform 20.2 in.
Length (Metric) 80.5 cm
Length (Metric) Platform 51.3 cm
Width (English) 51.9 in.
Width (English) Interior 40.4 in.
Width (English) Platform 36.6 in.
Width (Metric) 131.8 cm
Width (Metric) Interior 102.5 cm
Width (Metric) Platform 93 cm
Model CellXpert CS220
Display 7 in. Color Touchscreen
Humidity Range 20 to 85% (non-condensing)
Refrigerated No
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.