missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CELT-327

Product Code. 18075942
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18075942 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18075942 Supplier Tocris Bioscience™ Supplier No. 7954/10U

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective hA2B/A3 adenosine receptor fluorescent antagonist

CELT-327 is a potent and selective fluorescent hA2B/A3 adenosine receptor antagonist (Ki values are 35.6 nM and 45.7 nM at A2B and A3 receptors respectively). It shows dose-dependent membrane fluorescent labeling in HCT116 cells expressing A2B adenosine receptors, with rapid diffusion and homogeneous distribution of the fluorescence signal within the cell monolayer. In live HCT116 compact spheroids, CELT-327 displays homogeneous staining across the entire spheroid diameter, reaching deep spheroid regions. CELT-327 can be used as a fluorescent adenosine receptor probe for fluorescence binding assays, live cell imaging, flow cytometry, and fluorescence polarization assays. It is a suitable replacement for GPCR radiolabeling ligands. Excitation and emission maxima (λ) are 589 nm and 616 nm, respectively. Wavelengths are compatible with use as an acceptor dye in TR-FRET assays, together with the CoraFluor™ 1 (Cat. No. 7920) TR-FRET donor. Add 75 μL of assay buffer containing 1% of DMSO to obtain a 100 μM stock solution, and use 250 nM as a working concentration.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 μg
Target Fluorescent Receptor Probes
Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.