missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CENTD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24620SS
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
CENTD1 Polyclonal specifically detects CENTD1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Spezifikation
| CENTD1 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| ARAP2 | |
| A portion of amino acids 1670-1704 of human ARAP2 was used as the immunogen. | |
| 0.025 mg | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a, CRA_b, CRA_c, CRA_d and CRA_e), 93% homologous in rhesus monkey, 87% homologous in mouse and rat, 81% homologous in dog and 75% homologous in opossum. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 1.0-2.0 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml | |
| Arf and Rho GAP adapter protein 2, ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 2, centaurin, delta 1, Centaurin-delta-1, CENTD1arf-GAP with Rho-GAP domain, ANK repeat and PH domain-containing protein 2, cnt-d1, FLJ13675, FLJ44916, KIAA0580centaurin-delta-1, PARX, Protein PARX | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 116984 | |
| Human | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur