Description
- Save time with our strong and absorbent 2-ply paper that swiftly handles everyday wiping tasks.
- Effectively clean hands and lightly soiled surfaces with this multipurpose design.
- Users can take as much paper as they need, thanks to the unrestricted flow feature.
- Easy to use centrefeed dispenser that can be operated with just one hand.
- This paper can be used in the Tork™ Mini Centrefeed dispenser, which is a compact versatile solution for professional environments where both hand and surface wiping is required.
Specifikationer
Specifikationer
| Product Type | Wiping Paper |
| Length (English) | 81.91 yd. |
| Length (Metric) | 74.9 m |
| Width (English) | 7.64 in. |
| Width (Metric) | 19.4 cm |
| Material | Paper |
| Autoclavable | Not Autoclavable |
| Certifications/Compliance | FSC Chain-Of-Custody, OHSAS 18001, ISO 9001, EU Ecolabel, ISO 14001, FSC MIX Credit |
| Color | White |
| Disposable | Limited-use |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?