missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoraLite™ Plus 488-conjugated CYP2E1-Specific Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67263-100UL
This item is not returnable.
View return policy
Description
The CYP2E1-Specific antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human CYP2E1-Specific. This antibody recognizes mouse, rat, pig, human antigen. The CYP2E1-Specific antibody has been validated for the following applications: IF-P analysis. This antibody is coralite™ plus 488.Specifications
| CYP2E1-Specific | |
| Monoclonal | |
| 1000 μg/mL | |
| CPE1; CYP2E; P450-J; P450C2E | |
| Peptide | |
| 100 μL | |
| Primary | |
| Mouse, Rat, Pig, Human | |
| 50-57 kDa | |
| Liquid |
| Immunofluorescence | |
| 1F12G7 | |
| CoraLite Plus 488 | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 1571 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction