Learn More
CEP192 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28700-1-AP-20UL
Description
The CEP192 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human CEP192. This antibody recognizes human antigen. The CEP192 antibody has been validated for the following applications: IF, WB,ELISA analysis.
Specifications
| CEP192 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 55125 | |
| Liquid |
| Western Blot, Immunofluorescence, ELISA | |
| 1000 μg/mL | |
| Rabbit | |
| Primary | |
| Human |