Learn More
Invitrogen™ CEP63 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5113199
Product Code. 17101064
  • 461.00€ / 100µg
Description
CEP63 Polyclonal Antibody for Western Blot, IHC (P), ELISA
CEP63 is a protein with six coiled-coil domains. The protein is localized to the centrosome, a non-membranous organelle that functions as the major microtubule-organizing center in animal cells.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CEP63
Polyclonal
Unconjugated
CEP63
4921501M07; AL450317.13gm1; AW107703; CD20R; centrosomal protein 63; centrosomal protein 63kDa; centrosomal protein of 63 kDa; centrosome protein Cep63; CEP63; D9Mgc41; D9Mgc48e; ET2; LOW QUALITY PROTEIN: centrosomal protein of 63 kDa; SCKL6
Rabbit
Protein G
RUO
28135, 80254
-20°C or -80°C if preferred
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Q3UPP8, Q96MT8
CEP63
Recombinant Human Centrosomal protein of 63 kDa protein (1-495AA).
100 μg
Primary
Human, Mouse