missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CER1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA578286

Product Code. 16386984

  • 560.00€ / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Predicted Reactivity: Rhesus Monkey (89%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a cytokine member of the cysteine knot superfamily, characterized by nine conserved cysteines and a cysteine knot region. The cerberus-related cytokines, together with Dan and DRM/Gremlin, represent a group of bone morphogenetic protein antagonists that can bind directly to BMPs and inhibit their activity.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CER1
Polyclonal
Unconjugated
CER1
Cer1; cer-1; Cerberus; cerberus 1 homolog (Xenopus laevis); cerberus 1, cysteine knot superfamily, homolog; cerberus 1, DAN family BMP antagonist; cerberus homolog; cerberus-like protein; cerberus-related 1; cerberus-related protein; Cerl; Cer-l; Cerl1; Cerr1; DAN domain family member 4; DAND4; MGC119894; MGC119895; MGC96951; RGD1563046
Rabbit
Antigen affinity chromatography
RUO
9350
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
1 mg/mL
PBS with 20% glycerol and 0.01% thimerosal; pH 7
O95813
CER1
Full length human CER1 Recombinant protein.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.