Learn More
CERS2 Monoclonal Antibody (OTI3D9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809918
Description
LASS2 is a protein that has sequence similarity to yeast longevity assurance gene 1. Mutation or overexpression of the related gene in yeast has been shown to alter yeast lifespan. The human protein may play a role in the regulation of cell growth. This gene encodes a protein that has sequence similarity to yeast longevity assurance gene 1. Mutation or overexpression of the related gene in yeast has been shown to alter yeast lifespan. The human protein may play a role in the regulation of cell growth. Alternatively spliced transcript variants encoding the same protein have been described.Specifications
| CERS2 | |
| Monoclonal | |
| Unconjugated | |
| CERS2 | |
| L3, LASS2, SP260, TMSG1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 29956 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3D9 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96G23 | |
| CERS2 | |
| Human recombit protein fragment corresponding to amino acids 62-136 of human CERS2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.