missing translation for 'onlineSavingsMsg'
Learn More

Cesium chromate, 99+%, Thermo Scientific™

Product Code. 11495647
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11495647 10 g 10g
11405657 50 g 50g
11415657 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11495647 Supplier Thermo Scientific Alfa Aesar Supplier No. B23619.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13454-78-9
Molecular Formula CrCs2O4
Molecular Weight (g/mol) 381.80
MDL Number MFCD00016050
InChI Key BROHICCPQMHYFY-UHFFFAOYSA-N
Synonym cesium chromate, cesium chromate 99.9+%, dicesium dioxido dioxo chromium, dicesium chromate, cesium chromate cs2cro4, chromic acid h2cro4 , dicesium salt, caesium chromate, cro4.2cs, acmc-1bym2, dicaesium 1+ chromate 2-
PubChem CID 61613
IUPAC Name dicesium;dioxido(dioxo)chromium
SMILES [Cs+].[Cs+].[O-][Cr]([O-])(=O)=O

Specifications

Quantity 10 g
UN Number UN3087
Formula Weight 381.81
Percent Purity ≥99%
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Cesium chromate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.