missing translation for 'onlineSavingsMsg'
Learn More

Cesium Iodide, TRC

Product Code. 30355649
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Product Code. Quantity unitSize
30355649 250 mg 250mg
1 options
This item is not returnable. View return policy

Product Code. 30355649

Brand: Toronto Research Chemicals TRCC280018250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cesium Iodide
CAS 7789-17-5
Molecular Formula CsI
Quantity 250 mg
InChI Formula InChI=1S/Cs.HI/h;1H/q+1;/p-1
SMILES [I-].[Cs+]
IUPAC Name cesium;iodide
Molecular Weight (g/mol) 259.81
Formula Weight 259.8099
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.