Learn More
Cesium Trifluoroacetate (CsTFA), Cytiva
Product Code. 10744927 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10744927 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10744927 Supplier Life Science products Cytiva Supplier No. 17084702
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
  • Effective inhibitor of ribonucleases
  • Can separate RNA and DNA
  • Soluble in ethanol so nucleic acids can be recovered from the gradient by ethanol precipitation
For the separation and purification of nucleic acids, especially RNA, by isopycnic centrifugation
TRUSTED_SUSTAINABILITY
Specifications
Density 2.0 ± 0.05 g/mL
Synonym CsTFA, aqueous solution
Quantity 100 mL
Chemical Name or Material Cesium Trifluoroacetate Solution
Signal Word
  • Warning