missing translation for 'onlineSavingsMsg'
Learn More

Cesium Trifluoroacetate, TRC

Product Code. 30360491
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30360491 500 mg -
30351378 5 g 5g
30353139 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30360491 Supplier Toronto Research Chemicals Supplier No. TRCC280013500MG
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21907-50-6
Molecular Formula C2O2F3Cs
Molecular Weight (g/mol) 245.92
Synonym Acetic acid, trifluoro-, cesium salt (8CI,9CI)
IUPAC Name cesium;2,2,2-trifluoroacetate
SMILES [Cs+].[O-]C(=O)C(F)(F)F

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C2HF3O2.Cs/c3-2(4,5)1(6)7;/h(H,6,7);/q;+1/p-1
Formula Weight 245.8905
Chemical Name or Material Cesium Trifluoroacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.