missing translation for 'onlineSavingsMsg'
Learn More

Cetearyl Alcohol, TRC

Product Code. 30137800
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137800 5 g 5g
30137181 10 g 10g
30011685 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137800 Supplier Toronto Research Chemicals Supplier No. TRCC3648835G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67762-27-0
Molecular Formula C18H38O . C16H34O
Molecular Weight (g/mol) 512.93
Synonym Adol 63, Adol 65, Adol 66, Alcs., C16-18, Alfol 1618, Alfol 1618C, Alfol 30F, BTMS 350, Barolub LOH, C16-18 alcohols, C16-18 alcs., C16-18 fatty alcohols, CO 1618, CS 50, Cestopal 80M, Cetalol SCA, Cetanol K, Cetearyl alcohol, Cetostearyl alcohol, Cetylstearyl alcohol, Cire algonol CS, Cire deLanol ST, Conol 30OC, Crodacol 1618, Crodacol CS 50, Crodacol CS 90, Crodacol SCB, Crodamol CS 90, Cyclochem emulsion wax, Dehydag wax N, Ecorol 68/50P, Epal 1618, Hydrenol D, Hydrenol DV, Hydrenol MY, Hyfatol CS, Hyfatol CS 50, Hyfatol CS/EP, Kalcohl 68, Kalcohl 6850, Kalcohl 6870, Kalcohl 86, Kalcohl 8665, Kalcohl 8688, Kalcol 1618, Kalcol 6850, Kolliphor CS-A, Kolliphor CS-B, Kolliwax CSA 50, Kolliwax CSA 70, Lanette 20, Lanette D, Lanette O, Laurex CS, Mixture of cetyl and stearyl alcohols, NAA 45, NAA 46, Nafol 1618, Nafol 1618F, Radianol 1769, Rofanol P 50/55, Rofanol P 80/55, Rofanol P 80/85, Salim C 16/18, Sipol CS, Speziol C 16-18, Speziol D Pharma, Stenol 1618, Stenol PC, Surfol 1618, Tego Alkanol 1618
IUPAC Name heptadecan-1-ol
SMILES CCCCCCCCCCCCCCCCO.CCCCCCCCCCCCCCCCCCO

Specifications

Quantity 5 g
InChI Formula InChI=1S/C17H36O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18/h18H,2-17H2,1H3
Formula Weight 512.5532
Chemical Name or Material Cetearyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.