missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Cetrorelix acetate

Product Code. 30306094
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30306094 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30306094 Supplier Tocris Bioscience™ Supplier No. 8821/1
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Potent GnRH receptor antagonist

Cetrorelix acetate is a potent gonadotropin-releasing hormone (GnRH) receptor antagonist (KD = 0.202 nM, IC50 = 1.21 nM). Suppresses production of luteinizing hormone (LH) and follicle-stimulating hormone (FSH) from the pituitary gland, which inhibits ovulation. Exhibits antiproliferative effects and displays efficacy against hormone-sensitive cancers in vivo. Also exhibits anxiolytic and antidepressant activity in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Purity 95%
CAS 145672-81-7
Content And Storage Store at -20°C
Quantity 1 mg
Target Entry GnRH Antagonist
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.