missing translation for 'onlineSavingsMsg'
Learn More

Cetylpyridinium Chloride Monohydrate, Mikromol™

Product Code. 30049998
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30049998 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30049998 Supplier Mikromol Supplier No. MM1554.000250
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Cetylpyridinium Chloride Monohydrate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cetylpyridinium chloride monohydrate
CAS 6004-24-6
Analyte or Component Names Cetylpyridinium chloride monohydrate
Shipping Condition Room Temperature
Molecular Formula C21 H38 N . Cl . H2 O
Certifications/Compliance ISO 9001
Synonym Cetylpyridinium chloride, 1-hexadecylpyridinium chloride, Pyridinium, 1-hexadecyl-, chloride, hydrate (1:1:1), Pyridinium, 1-hexadecyl-, chloride, monohydrate (8CI,9CI), Cetylpyridinium chloride monohydrate, N-Cetylpyridinium chloride monohydrate, N-Hexadecylpyridinium chloride monohydrate
InChI Formula InChI=1S/C21H38N.ClH.H2O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-16-19-22-20-17-15-18-21-22;;/h15,17-18,20-21H,2-14,16,19H2,1H3;1H;1H2/q+1;;/p-1
SMILES O.[Cl-].CCCCCCCCCCCCCCCC[n+]1ccccc1
IUPAC Name 1-hexadecylpyridin-1-ium;chloride;hydrate
Molecular Weight (g/mol) 358
Quantity 250 mg
Formula Weight 357.2798 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.