missing translation for 'onlineSavingsMsg'
Learn More

Cetyltrimethyl Ammonium Bromide, Serva Electrophoresis™

Product Code. 11731917
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11731917 100 g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11731917 Supplier Serva Electrophoresis Supplier No. 1653004
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 57-09-0
Molecular Formula C19H42BrN
MDL Number MFCD00011772
Quantity 100 g
Synonym cetrimonium bromide, hexadecyltrimethylammonium bromide, cetyltrimethylammonium bromide, ctab, centimide, cetab, bromat, ctmab, cee dee, cetavlon
InChI Key LZZYPRNAOMGNLH-UHFFFAOYSA-M
SMILES [Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C
IUPAC Name hexadecyltrimethylazanium bromide
Molecular Weight (g/mol) 364.46
PubChem CID 5974
ChEBI CHEBI:3567
Physical Form Crystalline
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cetyltrimethylammonium-bromide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.