Learn More
Spectrum Labs™ CF-2 Fraction Collector
Product Code. 11943999
Click to view available options
Width (Metric):
28 cm
Unit Size:
Each
Product Code. 11943999
Brand: Spectrum Labs™ 124878
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Very durable, general-purpose fraction collector featuring a rugged drive and case. Spectrum Labs™ CF-2 Fraction Collector is sold complete and ready to use.
  • Stationary drop head
  • Tubes move under drop former, ensuring every drop is collected
  • Meets preparative LC and open column LC needs
  • Can collect up to 174 fractions in 13mm tubes or up to 116 fractions in 10mm tubes
  • Can collect directly into 17mm or 28mm scintillation vials
  • Cold-room compatible
  • CE marked
  • Complies with RoHS
Specifications
Depth (Metric) 40 cm
Height (Metric) 12 cm
Width (Metric) 28 cm
Description Includes: Drop counter, peak separator, tubing fittings. Does not include tube racks.
Electrical Requirements 230 VAC ±40 VAC, 20 VA, 45-65 Hz