Learn More
cGAS Mouse, anti-Human, Clone: 2G2C5, Proteintech
Mouse Monoclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | cGAS |
|---|---|
| Clone | 2G2C5 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
Description
The cGAS antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human cGAS. This antibody recognizes human antigen. The cGAS antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| cGAS | |
| 1000 μg/mL | |
| Liquid | |
| Human | |
| C6orf150, cGAS, h-cGAS, MB21D1 | |
| Primary |
| 2G2C5 | |
| Western Blot, ELISA | |
| Mouse | |
| 115004 | |
| IgG1 | |
| Store at -20°C. Stable for one year after shipment. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.