missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CGI-09 Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-50040APC
This item is not returnable.
View return policy
Description
CGI-09 Polyclonal antibody specifically detects CGI-09 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| CGI-09 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| CGI-09, GCD10, Gcd10p, KIAA1153, MGC5029, TRM6, tRNA (adenine-N(1)-)-methyltransferase non-catalytic subunit TRM6, tRNA methyltransferase 6 homolog (S. cerevisiae), tRNA(m1A58)-methyltransferase subunit TRM6, tRNA(m1A58)MTase subunit TRM6 | |
| The immunogen for this product maps to a region between residue 447 and 497 of human tRNA Methyltransferase 6 using the numbering given in entry NP_057023.2 (GeneID 51605). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| APC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 51605 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction