Learn More
CGRRF1 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16101-1-AP-20UL
 View more versions of this product
Product Code. 16875594
  • 145.00€ / 20µL
Description
CGRRF1 is able to inhibit growth in several cell lines.
Specifications
Specifications
CGRRF1
Polyclonal
Unconjugated
CGRRF1
CGR19, CGRRF1, RING finger protein 197, RNF197
Rabbit
Antigen Affinity Chromatography
RUO
10668, 116679
-20°C
Liquid
Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P97587, Q99675
CGRRF1
CGRRF1 Fusion Protein Ag8998
20 μL
Primary
Rat, Human