missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHCHD3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-21606
This item is not returnable.
View return policy
Description
CHCHD3 Polyclonal specifically detects CHCHD3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CHCHD3 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| coiled-coil-helix-coiled-coil-helix domain containing 3, FLJ20420, mitochondrial | |
| Rabbit | |
| 26 kDa | |
| 0.1 mL | |
| metabolism | |
| 54927 | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1mg/mL | |
| Western Blot 1:500-1:10000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| Q9NX63 | |
| CHCHD3 | |
| Recombinant protein encompassing a sequence within the center region of human CHCHD3. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse. Expected to cross react based on sequence identity: Pig (91%), Rat (83%), Bovine (90%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction