missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHD2 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12311-1-AP-20UL
This item is not returnable.
View return policy
Description
The CHD family of proteins is characterized by the presence of chromo (chromatin organization modifier) domains and SNF2-related helicase/ATPase domains. CHD proteins alter gene expression by modifying chromatin structure,thereby altering access of the transcriptional machinery to its chromosomal DNA template. Chromodomain helicase DNA binding protein 2, also known as CHD2, is a member of the SNF2/RAD54 helicase family of chromatin remodeling and DNA-binding proteins (CDH proteins). Consisting of 1,828 amino acids, CHD2 is expressed in the nucleus and contains two chromo domains, a helicase ATP-binding domain and a helicase C-terminal domain. CHD2 is suggested to play a critical role in development, hematopoiesis and tumor suppression. Mutations in the gene encoding CHD2 may be the cause of glomerulopathy, proteinuria and significantly impaired kidney function. CHD2 exists as two alternatively spliced isoforms.Specifications
| CHD2 | |
| Polyclonal | |
| Unconjugated | |
| CHD2 | |
| ATP dependent helicase CHD2, CHD 2, CHD2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1106, 308738 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O14647 | |
| CHD2 | |
| CHD2 Fusion Protein Ag2960 | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction