missing translation for 'onlineSavingsMsg'
Learn More

CHEMetrics CHEMets Dissolved Oxygen Test Kit Refill

Product Code. 11731044
Change view
Click to view available options
Range:
0 to 1 ppm
0 to 40 ppb and 0 to 100 ppb
1 to 12 ppm
Test Method:
Indigo Carmine
Rhodazine D
For Use With (Equipment):
C-7501 comparators
C-7512 comparators
C-7540 comparators
Unit Size:
Pack of 30
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range Test Method For Use With (Equipment) Unit Size
11731044 0 to 1 ppm Rhodazine D C-7501 comparators Pack of 30
11741044 1 to 12 ppm Indigo Carmine C-7512 comparators Pack of 30
11751044 0 to 40 ppb and 0 to 100 ppb Rhodazine D C-7540 comparators Pack of 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11731044 Supplier CHEMetrics Supplier No. R7501
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Provides results in two minutes or less

  • Designed for maximum simplicity and accuracy, Chemets 7 mm diameter ampules feature tapered, prescored tips with reagent vacuum-sealed inside
  • To perform a test, simply immerse Chemets ampule in the sample, snap off tip and the correct sample volume is quickly drawn inside the ampule
  • In two minutes or less, the reaction between the sample and reagent yields a color—which can then be compared to appropriate color standard for quantification of results
  • Comparators have two-year shelf life

Specifications

Analytes Dissolved Oxygen (DO)
For Use With (Equipment) C-7501 comparators
Includes 30 self-filling CHEMets ampoules to refill kit K-7501
No. of Tests 30
Range 0 to 1 ppm
Test Method Rhodazine D
Test Time 2 min.
Type CHEMets Dissolved Oxygen Refill
compliance-icons compliance-icons
Product Identifier
  • Oxygen, dissolved
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/physician/first aider.
  • P363-Wash contaminated clothing before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.