Learn More
CHEMetrics CHEMets Sulfide Visual Water Test Kit
Product Code. 11721014
Change view
Click to view available options
No. of Tests:
30
Unit Size:
Pack of 30
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Tests unitSize
11721014 30 Pack of 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11721014 Supplier CHEMetrics Supplier No. K9510
Includes
REMOVE
Provides results in two minutes or less
Provide results in two minutes or less
Comes in a plastic case and contains everything needed to perform 30 tests: Refill, Low & High Range Comparators, Activator Solution, 25 mL sample cup and instructions.
Specifications
Analytes Sulfide
Includes Plastic case, Refill, Low and High range comparators, Activator Solution, 25 mL sample cup, instructions
No. of Tests 30
Range 0 to 1 ppm and 1 to 10 ppm
Test Method Methylene Blue
Test Time 5 min.
Type CHEMets Visual Water Testing Kit
compliance-icons compliance-icons
Product Identifier
  • CHEMets sulphide test 0-10PPM
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P234-Keep only in original container.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P363-Wash contaminated clothing before reuse.
  • P405-Store locked up.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: hydrochloric acid, ferric chloride, hexahydrate, isopropanol, N,N-dimethyl-p-phenylenediamine oxalat