missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHEMetrics CHEMets Chlorine Test Kit Refill
Description
- Designed for maximum simplicity and accuracy, Chemets 7 mm diameter ampules feature tapered, prescored tips with reagent vacuum-sealed inside
- To perform a test, simply immerse Chemets ampule in the sample, snap off tip and the correct sample volume is quickly drawn inside the ampule
- In two minutes or less, the reaction between the sample and reagent yields a color—which can then be compared to appropriate color standard for quantification of results
- Comparators have two-year shelf life
Specifications
Specifications
| Analytes | Chlorine |
| For Use With (Equipment) | Chlorine (free and total) testing kit |
| Includes | 30 ampoules |
| Range | 0 to 1 ppm and 1 to 5 ppm |
| Test Method | DDPD |
| Test Time | 2 min. |
| Type | CHEMets Refill |
Safety and Handling
Product Identifier
- Chlorine CHEMets & ULR CHEMets Refills
- Danger
- Acute toxicity Category 4
- Carcinogenicity Category 1B
- Specific target organ toxicity Category 2
- H317-May cause an allergic skin reaction.
- H302-Harmful if swallowed.
- H350-May cause cancer.
- H371-May cause damage to organs.
- EUH208-Contains (EDTA disodium saltdihydrate, formaldehyde). May produce an allergic reaction.
- P201-Obtain special instructions before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P270-Do not eat, drink or smoke when using this product.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P405-Store locked up.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: methanol, EDTA disodium salt dihydrate,sulfuric acid, formaldehyde
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?