missing translation for 'onlineSavingsMsg'
Learn More

CHEMetrics CHEMets Chlorine Test Kit Refill

Product Code. 11777433
Change view
Click to view available options
Range:
0 to 1 ppm and 1 to 5 ppm
0.1 to 2 ppm
Test Method:
DDPD
DPD
Unit Size:
Pack of 30
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range Test Method Unit Size
11777433 0.1 to 2 ppm DPD Pack of 30
11771024 0 to 1 ppm and 1 to 5 ppm DDPD Pack of 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11777433 Supplier CHEMetrics Supplier No. R2500
Power Your Renewable Energy Research
Use promo code 28029
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Provides results in two minutes or less

  • Designed for maximum simplicity and accuracy, Chemets 7 mm diameter ampules feature tapered, prescored tips with reagent vacuum-sealed inside
  • To perform a test, simply immerse Chemets ampule in the sample, snap off tip and the correct sample volume is quickly drawn inside the ampule
  • In two minutes or less, the reaction between the sample and reagent yields a color—which can then be compared to appropriate color standard for quantification of results
  • Comparators have two-year shelf life

Specifications

Analytes Chlorine
For Use With (Equipment) Chlorine (free and total) testing kit
Includes 30 ampoules
Range 0.1 to 2 ppm
Test Method DPD
Test Time 2 min.
Type CHEMets Refill
compliance-icons compliance-icons
Product Identifier
  • dissolved oxygen vacu-vials ampoules and chemets refill
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: diethylene glycol, potassium hydrogen phthalate, hydrochloric acid, EDTA disodium salt dihydrate, C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.