Learn More
Hanna Instruments™ Chemical Test Kit
Product Code. 11763243
Change view
Click to view available options
For Use With (Application):
Free Chlorine LR/MR Determination
Free and Total Chlorine Determination
Iron (Fe2+ and Fe3+) Determination
Nitrite Determination
Phenols Determination
Phosphate Determination
Sulfite Determination
Total Chlorine Determination
Total Hardness Determination
Unit Size:
Each
Pack of 100
Pack of 50
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
11763243 Sulfite Determination Pack of 100
10504784 Iron (Fe2+ and Fe3+) Determination Each
11692300 Free Chlorine LR/MR Determination Each
11612310 Free and Total Chlorine Determination Each
11676770 Nitrite Determination Each
11713243 Total Chlorine Determination Pack of 100
11773233 Phosphate Determination Pack of 50
11783233 Total Hardness Determination Pack of 50
11703253 Phenols Determination Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 missing translation for 'options'
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 11763243 missing translation for 'mfr' Hanna Instruments™ missing translation for 'supplierNo' HI3822
  • ONLINE EXCLUSIVE PRICE
    182.70€
    161.08€ / Pack of 100
    Save 21.62€
    11% Off
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Includes all of the reagents and equipment necessary to perform manual water testing. Hanna Instruments™ Chemical Test Kit features pre-made, pre-measured reagents for fast, accurate determinations with high-resolution readings based on accepted titrations methods.
These chemical test kits use high quality reagents that are manufactured in Hanna's state-of-the-art facility and are clearly marked with the lot number and expiration date on each packet/bottle for traceability.
  • Complete setup: All required materials are included, such as beakers, syringes, and premeasured reagent bottles and packets
  • High resolution readings for fast, accurate determinations
  • Replacement reagents available: No need to buy a new kit when reagents are exhausted
Specifications
For Use With (Application) Sulfite Determination
For Use With (Equipment) Water samples
Includes 30mL sulfamic acid solution, 30mL EDTA reagent, 15mL sulfuric acid solution, 10mL starch indicator, 120mL titrant solution, 20mL calibrated vessel, 50mL calibrated vessel, and calibrated syringe with tip
No. of Tests 110
Quantity 1 Kit
Range 0 to 20mg/L, 0 to 200mg/L
Sufficient For Two ranges
Test Method Iodometric
Type Sulfite Test Kit
compliance-icons compliance-icons compliance-icons
Product Identifier
  • X100 TEST SULFITE
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H372-Causes damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362-Take off contaminated clothing.
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contain: potassium iodide, sulphamic acid, sulphuric acid