missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Chemical Test Kit

Product Code. 11783233
Change view
Click to view available options
For Use With (Application):
Free Chlorine LR/MR Determination
Free and Total Chlorine Determination
Iron (Fe2+ and Fe3+) Determination
Nitrite Determination
Phenols Determination
Phosphate Determination
Sulfite Determination
Total Chlorine Determination
Total Hardness Determination
Unit Size:
Each
Pack of 100
Pack of 50
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
11783233 Total Hardness Determination Pack of 50
10504784 Iron (Fe2+ and Fe3+) Determination Each
11692300 Free Chlorine LR/MR Determination Each
11612310 Free and Total Chlorine Determination Each
11676770 Nitrite Determination Each
11713243 Total Chlorine Determination Pack of 100
11763243 Sulfite Determination Pack of 100
11773233 Phosphate Determination Pack of 50
11703253 Phenols Determination Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 11783233 Supplier Hanna Instruments™ Supplier No. HI3840
  • ONLINE EXCLUSIVE PRICE
    33.02€
    19.76€ / Pack of 50
    Save 13.26€
    40% Off
View all promotions
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes all of the reagents and equipment necessary to perform manual water testing. Hanna Instruments™ Chemical Test Kit features pre-made, pre-measured reagents for fast, accurate determinations with high-resolution readings based on accepted titrations methods.

These chemical test kits use high quality reagents that are manufactured in Hanna's state-of-the-art facility and are clearly marked with the lot number and expiration date on each packet/bottle for traceability.

  • Complete setup: All required materials are included, such as beakers, syringes, and premeasured reagent bottles and packets
  • High resolution readings for fast, accurate determinations
  • Replacement reagents available: No need to buy a new kit when reagents are exhausted

Specifications

Analytes EDTA
For Use With (Application) Total Hardness Determination
For Use With (Equipment) Water samples
Includes 30mL hardness LR reagent and 50mL calibrated vessel
No. of Tests 50
Quantity 1 Kit
Range 0 to 150mg/L
Sufficient For Low range determination
Test Method Titration
Type Total Hardness Low Range Test Kit
compliance-icons
Product Identifier
  • HARDNESS KIT LOW RANGE
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: edta tetrasodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.