missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chitinase 3-like 1 Antibody (CL12241), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21212-100ul
This item is not returnable.
View return policy
Description
Chitinase 3-like 1 Monoclonal antibody specifically detects Chitinase 3-like 1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Chitinase 3-like 1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL12241 | |
| Western Blot 1 ug/ml, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| AW208766, Brp39, Chi3l1, gp39 | |
| This antibody was developed against a Synthetic Peptide of P36222. Actual immunogen sequence is proprietary. | |
| 100 μg | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 1116 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction