missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chitinase 3-like 1/YKL-40 Antibody (CL12314) [Alexa Fluor™ 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-15132AF594
This item is not returnable.
View return policy
Description
Chitinase 3-like 1/YKL-40 Monoclonal antibody specifically detects Chitinase 3-like 1/YKL-40 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Chitinase 3-like 1/YKL-40 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| AW208766, Brp39, Chi3l1, gp39, YKL-40 | |
| This antibody was developed using a synthetic peptide derived from P36222, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 1116 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL12314 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction