missing translation for 'onlineSavingsMsg'
Learn More

alpha-Chloralose, 98+%, (max. 20% beta-anomer)

Product Code. 10407920 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10407920 100 g 100g
10655962 25 g 25g
10656152 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10407920 Supplier Thermo Scientific Acros Supplier No. 147791000
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

alpha-Chloralose, CAS #15879-93-3 is a true anesthetic or an immobilizing agent with sedative-hypnotic properties. | CAS: 15879-93-3 | C8H11Cl3O6 | 309.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • alpha-Chloralose is an anesthetic and an immobilizing agent
  • It involves autonomic reflexes

Applications

  • alpha-Chloralose is an anesthetic with sedative-hypnotic properties
  • It has been used in laboratory to anesthetize certain animals
  • It can be used to immobilize certain animals in the laboratory setting
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15879-93-3
Molecular Formula C8H11Cl3O6
Molecular Weight (g/mol) 309.52
MDL Number MFCD00005542
InChI Key OJYGBLRPYBAHRT-OPKHMCHVSA-N
Synonym a-chloralose, 1-2r,5r,6s,6ar-6-hydroxy-2-trichloromethyl-3a,5,6,6a-tetrahydrofuro 2,3-d 1,3 dioxol-5-yl ethane-1,2-diol, r-1-2r,3ar,5r,6s,6ar-6-hydroxy-2-trichloromethyl tetrahydrofuro 2,3-d 1,3 dioxol-5-yl ethane-1,2-diol
PubChem CID 7057995
IUPAC Name (1R)-1-[(2R,3aR,5R,6S,6aR)-6-hydroxy-2-(trichloromethyl)-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxol-5-yl]ethane-1,2-diol
SMILES OCC(O)[C@H]1OC2O[C@@H](O[C@@H]2[C@H]1O)C(Cl)(Cl)Cl

Specifications

Melting Point 176°C to 182°C
CAS Min % 96.0
Color White to Yellow
CAS Max % 100.0
Quantity 100 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Powder
Assay Percent Range 97%
Merck Index 15, 2072
Formula Weight 309.52
Percent Purity ≥96.0%
Chemical Name or Material α-Chloralose, 97%
Show More Show Less
compliance-icons
Product Identifier
  • a-Chloralose
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.