missing translation for 'onlineSavingsMsg'
Learn More

Chlordiazepoxide 1.0 mg/ml in Methanol, LoGiCal

Product Code. 30460909
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30460909 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30460909 Supplier LoGiCal Supplier No. LGCAMP0028.0801

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chlordiazepoxide
CAS 58-25-3
Analyte or Component Names Chlordiazepoxide
Shipping Condition Room Temperature
Molecular Formula C16 H14 Cl N3 O
Certifications/Compliance ISO 17034
Synonym 7-chloro-N-methyl-5-phenyl-3H-1,4-Benzodiazepin-2-amine 4-oxide, 7-Chloro-2-(methylamino)-5-phenyl-3H-1,4-Benzodiazepine 4-oxide, 7-Chloro-2-(methylamino)-5-phenyl-3H-1,4-benzodiazepine 4-oxide, Abboxide, Balance, Balance (pharmaceutical), CDO, Chlordiazepoxid, Chlordiazepoxide, Chlorodiazepoxide, Chlozepid, Clopoxide, Control, Decacil, Eden, Elenium, Helogaphen, Ifibrium, Kalmocaps, Librelease, Librinin, Libritabs, Menrium, Mesural, Methaminodiazepoxide, Mildmen, Multum, Napoton, Psicosan, Radepur, Risolid, Silibrin, Tropium, Viopsicol
InChI Formula InChI=1S/C16H14ClN3O/c1-18-15-10-20(21)16(11-5-3-2-4-6-11)13-9-12(17)7-8-14(13)19-15/h2-9H,10H2,1H3,(H,18,19)
Solution Type Single Solution
SMILES CNC1=Nc2ccc(Cl)cc2C(=[N+]([O-])C1)c3ccccc3
IUPAC Name 7-chloro-N-methyl-4-oxido-5-phenyl-3H-1,4-benzodiazepin-4-ium-2-amine
Solvent or Matrix Methanol
Industry Type Clinical Testing
Molecular Weight (g/mol) 299.75
Quantity 1 mL
Formula Weight 299.0825 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.