Learn More
Sherwood Scientific™ M926S Chloride Analyzer
Product Code. 12876813
Change view
Click to view available options
Voltage:
100/240 V
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage Unit Size
12876813 100/240 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 12876813 Supplier Sherwood Scientific™ Supplier No. 92600109
  • ONLINE EXCLUSIVE PRICE
    7050.00€
    6322.85€ / Pack of 1
    Save 727.15€
    10% Off
Estimated Shipment: 26-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
Analyze chloride content via reliable colorimetric titration methodology. Sherwood Scientific™ M926S Chloride Analyzer is an ergonomically designed and microprocessor-based instrument.
  • Colorimetric titration method where silver ions are generated in-situ and react with chloride ions.
  • Accurate, proven method for chloride and salt concentration.
  • Wide range of applications including those in clinical (Model 926S) and food industries.
  • Very easy to use.
  • Equipped with RS232 serial and USB outputs which enable them to be linked to a suitable printer or to be interfaced to a PC hosting Active Salt software.
  • Active Salt package can collect data directly from a balance with RS232 output together with the chloride analyser results.
  • Software may be configured by the operator to suit their particular laboratory requirements.
  • DataGate LIMS and auto reading of balance and barcode.
  • No transcription errors
  • No time wasted doing manual calculations.
  • No need to achieve specific sample and diluent weights.
  • Automatic data storage and analysis report generation.
  • End point precisely detected.
  • No reagent needing recalibration.
  • Economical per test.
Specifications
Sample Size 20 μL to 100μL
Includes Printer cable, acid buffer, standard, 3 anodes, set of electrodes, 2 marked beakers, electrode cleaning polish, stirrer and manual
Voltage 100/240 V